Butyl acetate ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate

434620025 434621000 434620010
TS434620025EA 225 USD
TS434620025 TS434621000 TS434620010
Butyl acetate ACS
Butyl acetate
Formula: CH₃COO(CH₂)₃CH₃
MW: 116.16 g/mol
Boiling Pt: 126
Melting Pt: -76
Density: 0.88 g/cm³
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00009445
CAS Number: 123-86-4
UN: 1123
ADR: 3 III
Merck Index: 13,01534

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.1 % (K.F.)
GC ≤0.2 % (n-Butyl alcohol)
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 Passes test
Titrable acid ≤0.0016 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR