Methyl ethyl ketone, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-ButanoneMEKButan-2-oneEK 500 AdditiveButanoneEthyl methyl ketone

444170050
TS444170050EA 69.4 USD
TS444170050
Methyl ethyl ketone, Technical Grade
Methyl ethyl ketone
Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: –87 °C
Density: 0.806 g/cm³
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
UN: 1193
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.2 %
GC ≥99.0 %
Color scale ≤10 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR