Methyl ethyl ketone 99+% for spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-ButanoneMEKButan-2-oneEK 500 AdditiveButanoneEthyl methyl ketone

213015000 213010025
TS213015000EA 76.5 USD
TS213015000 TS213010025
Methyl ethyl ketone 99+% for spectroscopy
Methyl ethyl ketone
Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79.6 °C (1013 hPa)
Melting Pt: –87 °C
Density: 0.806 g/cm³
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,II
REACH: 01-2119457290-43

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 %
GC ≥99.0 %
UV 1 cm cell vs water
UV at 330 nm A: ≤1
UV at 340 nm A: ≤0.1
UV at 350 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR