Methanol-[D₄] 99.8 atom % D, AcroSeal™ for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Carbinol-D4
, Methyl alcohol-D4
, Methanol-[²H₄]
, Methyl-D3 alcohol-D
, Wood alcohol-D4
, Methanol-d4
, Hydroxy methane-D4
, Tetradeuteromethanol
426951000
TS426951000EA
886
USD
TS426951000
Methanol-[D₄] 99.8 atom % D, AcroSeal™ for NMR spectroscopy
Methanol-[D4]
Formula:
D₃COD MW: 36.01 g/mol Boiling Pt: 65.3 °C (1013 hPa) Melting Pt: –99 °C Density: 0.888 g/cm³ (20 °C) Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044637 CAS Number: 811-98-3 UN: 1230 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.03 % |
D-Enrichment | ≥99.75 % |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...