Dichloromethane-[D₂] 99.6+ atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dichloromethane-[²H₂]Dideuteromethylene chlorideMethylene chloride-D2Methylene chloride-d2Dichloromethane-d2

217350100
TS217350100EA 260 USD
TS217350100
Dichloromethane-[D₂] 99.6+ atom % D for NMR spectroscopy
Dichloromethane-[D2]

Packaged in 1.00 ml ampoules

Formula: CD₂Cl₂
MW: 86.92 g/mol
Boiling Pt: 40 °C (1013 hPa)
Melting Pt: –97 °C
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000882
CAS Number: 1665-00-5
EINECS: 216-776-0
UN: 1593
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.04 % (Coulometric)
D-Enrichment ≥99.60 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR