Acetonitrile-[D₃] 99.9 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetonitrile-d3Cyanomethane-D3Ethannitrile-D3Methyl-d3 cyanideAcetonitrile-[²H₃]Methyl cyanide-D3Trideuteroacetonitrile

320650075
TS320650075EA 216 USD
TS320650075
Acetonitrile-[D₃] 99.9 atom % D for NMR spectroscopy
Acetonitrile-[D3]

Packaged in 0.75 ml ampoules

Formula: D₃CCN
MW: 44.03 g/mol
Boiling Pt: 80.7 °C (1013 hPa)
Melting Pt: –46 °C
Density: 0.844 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001881
CAS Number: 2206-26-0
EINECS: 218-616-5
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.030 % (Coulometric)
D-Enrichment ≥99.85 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR