AG 1478 >98% (by TLC, NMR)

Supplier: Sino Biological

E10-900B-05 E10-900B-25
77556-898EA 121 USD
77556-898 77556-900
AG 1478 >98% (by TLC, NMR)
AG 1478
Melting Pt: 177...178 °C
Storage Temperature: Freezer
CAS Number: 175178-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR