LH-846 >98% (by TLC, NMR)

Supplier: Sino Biological

C67-901-05 C67-901-25
77555-760EA 83 USD
77555-760 77555-762
LH-846 >98% (by TLC, NMR)
LH-846
Formula: C₁₆H₁₃ClN₂OS
Storage Temperature: Ambient
MDL Number: MFCD03786038
CAS Number: 639052-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR