MLS-573151 >98% (by TLC, NMR)

Supplier: Sino Biological

C08-909-05 C08-909-25
77555-080EA 155 USD
77555-080 77555-082
MLS-573151 >98% (by TLC, NMR)
MLS-573151
Formula: C₂₁H₁₉N₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD00358662
CAS Number: 10179-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR