LFM-A13 98% (by TLC, NMR)
Supplier: Sino Biological
|
|
B10-901-05
B10-901-25
77554-820EA
125
USD
77554-820
77554-822
LFM-A13 98% (by TLC, NMR)
LFM-A13
Formula:
C₁₁H₈Br₂N₂O₂ Melting Pt: 145...146 °C Storage Temperature: Freezer |
MDL Number:
MFCD02179204 CAS Number: 62004-35-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...