Dabrafenib >98% (by TLC, NMR)

Supplier: Sino Biological
Warning

Synonyms: N-(3-(5-(2-Aminopyrimidin-4-yl)-2-(tert-butyl)thiazol-4-yl)-2-fluorophenyl)-2,6-difluorobenzenesulphonamide, N-(3-(5-(2-Aminopyrimidin-4-yl)-2-(tert-butyl)thiazol-4-yl)-2-fluorophenyl)-2,6-difluorobenzenesulfonamide

B08-901-05 B08-901-25
77554-758EA 61 USD
77554-758 77554-760
Dabrafenib >98% (by TLC, NMR)
Dabrafenib
Formula: C₂₃H₂₀F₃N₅O₂S₂
Storage Temperature: Freezer
MDL Number: MFCD17215684
CAS Number: 1195765-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR