Biopreservation Cell Freezing Media and Antigens, Biopreservation Media and Cryopreservation Media
Supplier: Charles River Laboratories Cell Solutions
BPM101204 BPM210202 BPM210102 BPM205202 BPM101104 BPM101102 BPM210373 BPM202102 BPM205102
77529-680EA 86.92 USD
77529-680 77529-670 77529-672 77529-664 77529-674 77529-666 77529-676 77529-668 77529-678
Biopreservation Cell Freezing Media and Antigens, Biopreservation Media and Cryopreservation Media
Cell Freezing Media
BioLife Solutions and Charles River have partnered to provide CGMP-compliant biopreservation media products. This media significantly extends stability and improves post-preservation viability and function of biologic source material, intermediates, and final products.
Antigen products are proteins and viral lysates that stimulate in vitro immune responses. Various antigens are offered that stimulate immune cells, all of which are tested for bioactivity using PBMCs.
Biopreservation media and cryopreservation media: Biopreservation is used to suppress the biochemical process of aging and preserve those physiological functions, which lend cells and tissue their scientific and therapeutic value. Optimized cell freezing media can preserve cell viability and function such that it is equivalent to or, in some circumstances, better than freshly isolated products depending on the shipping logistics.
CryoStor® proprietary cryopreservation media products are pre-formulated with USP-grade DMSO and are intended for cryopreservation of biological materials at –70 to +196 °C.
HypoThermosol® biopreservation media is designed to provide maximum storage and shipping stability for biologics at 2 to 8 °C.
Delivery information: Cold packs, ambient temperature 2 to 8 °C or 15 to 25 °C.
Packaging: Glass vials or plastic bottles/bags as indicated by product type.
Caution: For research use only. Store in 2 to 8 °C but can ship at ambient temperature.