HemaPrime™ Antigen-Specific T Cells, Cryopreserved
Supplier: Charles River Laboratories Cell Solutions
HemaPrime™
ASTC-1049 ASTC-1083 ASTC-1093 ASTC-1022 ASTC-1048 ASTC-1092 ASTC-1122 ASTC-1089 ASTC-1072 ASTC-1123 ASTC-1125 ASTC-1126 ASTC-1084 ASTC-1099 ASTC-1127 ASTC-1039 ASTC-1142
77529-626EA 1074.15 USD
77529-626 77529-614 77529-636 77529-606 77529-628 77529-616 77529-638 77529-608 77529-618 77529-620 77529-630 77529-622 77529-610 77529-632 77529-624 77529-612 77529-634
HemaPrime™ Antigen-Specific T Cells, Cryopreserved
Blood
Peripheral blood cells are collected from healthy donors by direct collection or by using the Spectra Optia® Apheresis system. Cell-rich leukapheresis product is extracted using continuous flow centrifugal technology directly into the collection bag.
Purification, isolation, and cryopreservation of cells are performed on-site immediately following collection utilizing best-in-class instruments, biopreservation media, and standardized protocols. The rigorous operational controls mitigate any potential loss while ensuring viable and functional cells post-thaw, guaranteeing that customers receive consistent high-quality cells.
Antigen-specific T cells: Antigen-specific T cells (ASTCs) can recognize a specific peptide presented by antigen-presenting cells. These specific peptides can be derived from viruses, bacteria, and cancer cells. However, frequency of these ASTCs is very low among circulating T cells, making them difficult, if not impossible, to isolate. This impedes the ability to study how they function during induction of immune responses. Thus, ASTCs provide a unique tool for studying the activation and specificity of the immune system for early-stage research and discovery, as well as assays designed at measuring the ability of T cells to kill the target cells.
Delivery information: Dry ice or liquid nitrogen dewar.
Packaging: 1 ml cryovial.
Caution: For research use only. Store in Vapor Phase LN₂ until ready to use.