Water Soluble Paper Label Tapes for BradyJet™ J7300 Color Label Printer
Supplier: Brady Worldwide
176583 170460 170461 170462 176605
77580-962RL 65.19 USD
77580-962 77581-114 77580-908 77580-952 77581-060
Water Soluble Paper Label Tapes for BradyJet™ J7300 Color Label Printer
Labels
Choose water soluble paper labels for complete label and adhesive removal. Ideal for laboratories, temporary identification needs, and work-in-process product flows.
  • Dissolvable acrylic adhesive
  • Matte finish
  • Removable, water dissolvable material
  • Service temperature range: −94 to 212 °F
  • RoHS compliant to 2011/65/EU (Pass 2015/863)
Label and adhesive completely removes in 30 seconds using warm water. Inkjet receptive labels resist abrasions, humidity, and temperatures from −94 to 212 °F (−70 to 100 °C). Dissolvable acrylic adhesive is ideal for labs, asset tracking, and temporary ID. Built-in LabelSense™ technology enables fast cartridge changeovers and automatic label setup to save time, eliminate tasks, and reduce label waste.
For use with the BradyJet™ J7300 Color Label Printer. Ink cartridges are required to print on this material (use part numbers P-CMY-1 and P-K-1).