AlphaTec® 38-001 Butyl Gloves
Supplier: Ansell Healthcare
38001PP070 38001PP080 38001PP090 38001PP100 38001PP110
77322-828CS 997.92 USD
77322-828 77322-830 77322-832 77322-834 77322-836
AlphaTec® 38-001 Butyl Gloves
Gloves
AlphaTec® 38-001 butyl gloves offer superior protection against highly aggressive chemicals, including aldehydes, ketones, esters, and concentrated mineral acids.
  • Unique butyl formulation for a soft feel with a raised diamond pattern
  • Extended forearm protection
  • Excellent dry and wet grip to enhance dexterity and reduce hand fatigue
  • Wide cuff to allow donning over protection clothing
Designed for comfort, AlphaTec® 38-001 industrial gloves feature a unique butyl formulation to facilatate a soft feel, protecting hands while facilitating donning and doffing. The ultra-soft butyl formulation maximizes flexibility and dexterity, with a raised diamond pattern that enhances grip and ensures safer handling.
AlphaTec® 38-001 industrial gloves are recommended for use in automotive, life science, and chemical industries.
Certifications: EN ISO 21420:2020, EN ISO 373-1:2016 Type A, EN ISO 374-5:2016.
Packaging: Gloves are packaged 1 pair per bag, 36 bags per carton.