HyFlex® 11-591 Industrial Gloves, Vendor Pack
Supplier: Ansell Healthcare
11591060-VEND 11591070-VEND 11591090-VEND 11591100-VEND 11591110-VEND 11591120-VEND 11591080-VEND
77322-814CS 2021.44 USD
77322-814 77322-816 77322-820 77322-822 77322-824 77322-826 77322-818
HyFlex® 11-591 Industrial Gloves, Vendor Pack
Gloves
HyFlex® 11-591 industrial gloves provide enhanced comfort, with a 20% lighter design that allows extended use on varied tasks.
  • Ultra-lightweight design
  • 12 times greater cut resistance for reliable hand protection
  • Palm dipped, thumbcrotch reinforcement
  • Silicone-free and anti-static
HyFlex® 11-591 industrial gloves offer 12 times greater cut resistance thanks to the Next Generation single HPPE yarn developed with INTERCEPT™ Cut Resistance Technology for reliable hand protection. Featuring a 20% lighter design, these gloves provide enhanced comfort, and allow workers to perform varied tasks while keeping hands protected throughout extended use. Improved FORTIX™ Technology delivers up to 20% more abrasion resistance for longer-lasting handling in abrasive conditions. EN 407 contact heat level 1 to protect hands while handling hot parts.
HyFlex® 11-591 industrial gloves are recommended for use in automotive, glass manufacturing, heavy manufacturing, metal fabrication, machinery, equipment, and pulp/paper industries.
Certifications: ANSI Cut A9. ANSI ABR 5. EN 388:2016 +A1:2018, EN407:2020, EN ISO 21420:2020. Food Contact approved. Category II.
Packaging: Gloves are packaged 12 pairs per paper band bundle, 12 paper band bundles per carton.