Ward's® Hotplate Stirrer
Supplier: Ward's
Ward's®
MHS-800
470356-912EA 299 USD
470356-912
Ward's® Hotplate Stirrer
Stirrers Magnetic Hotplate Stirrers
Economical hotplate stirrer featuring advanced microprocessor technology to control both temperature and speed.
  • Chemical resistant ceramic top for easy cleaning and ideal sample visibility
  • 1.5 L stirring capacity
  • 13.3 cm (5.25") round platform
Control heat and speed accurately and quickly using the simple controls. Stirring starts safely with the unit's soft start feature to minimize splashing or irregular mixing.
When operating above 50 ℃ a safety LED lamp illuminates to indicate 'Hot' and remains lit until the unit has cooled.
Unit includes a detachable electric cord and a magnetic stirring bar. An optional support rod can be mounted to the threaded mounting port on the back.