Brady® Self-Laminating Vinyl Wrap-Around Labels
Supplier: Brady Worldwide
170752 170753 170754 170755 170756 170757 170758 170759 170760 170761 170863 170864 170865 170866 170867 170868 170869 170870 170740 170741 170742 170743 170744 170745 170746 170747 170748 170749 170750 170751 170861 170862
76795-802EA 64.87 USD
76795-802 76795-970 76795-776 76795-882 76796-202 76795-998 76795-850 76795-912 76795-854 76796-172 76796-182 76795-890 76796-198 76796-122 76795-834 76795-770 76796-204 76795-814 76795-718 76795-964 76796-030 76795-894 76795-778 76796-130 76795-752 76796-028 76796-092 76795-724 76796-006 76795-798 76795-756 76795-930
Brady® Self-Laminating Vinyl Wrap-Around Labels
Labels
Flexible labels are designed for tightly-curved surfaces such as wires and cables.
  • Permanent acrylic adhesive
  • Matte finish
  • Flame-retardant, self-laminating material
  • Recommended ribbon: R4300 (94000-380)
Durable labels are self-extinguishing and resistant to abrasion, solvents, oil, grease, weather, and extreme temperatures. Made with a permanent acrylic adhesive that works great on smooth, rough, powder-coated, and irregular surfaces. A clear, non-printable overlaminate provides increased protection.
Certifications: UL recognised to UL969 Labelling and Marking Standard when printed with the Brady series R4300 and R6200 ribbons. See UL files MH17154 for specific details. Both UL recognised B-427 labels and non-approved labels are available depending on customer requirements.