Brady® Destructible Tamper Evident Matte Vinyl Labels
Supplier: Brady Worldwide
170856 170857
76795-988EA 82.8 USD
76795-988 76796-156
Brady® Destructible Tamper Evident Matte Vinyl Labels
Labels
Specialized labels are designed for use in asset tracking, rating plates, product security, and tamper identification.
  • Permanent acrylic adhesive
  • Matte finish
  • Abrasion-Resistant, humidity-resistant, tamper-resistant, UV-resistant, weather-resistant material
  • Recommended ribbon: R6000HF
Tamper-resistant labels are designed to fracture to show signs of product tampering and prevent one-piece removal. The tough, permanent adhesive bonds tightly to almost any surface.
Certifications: RoHS compliant to 2011/65/EU (Pass 2015/863).