Atellica® VTR Syringes
Supplier: Siemens Healthineers
11561350 11561351 11561352 11561340 11561341 11561342 11561343 11561344 11561345 11561346 11561347 11561348 11561349
77553-722BX 447.23 USD
77553-722 77553-724 77553-726 77553-702 77553-704 77553-706 77553-708 77553-710 77553-712 77553-714 77553-716 77553-718 77553-720
Atellica® VTR Syringes
Syringes Syringes, Medical
The Atellica® VTR Arterial blood sampler syringes respond to the need for rapid, accurate blood collection and sample integrity. It is design reduces the potential for pre-analytical errors, featuring a dry, balanced, lithium-zinc heparin formulation that prevents coagulation with virtually no impact on five main electrolytes. These benefits are not affected by whole-blood samples with low protein content.
  • Feature ABG SafetyTip™ needlestick protection technology to help prevent accidental needlestick injuries
  • Easy to learn and simple to use single-handed safety activation
  • Provides optimum visualization of the insertion site
  • The patented dual-locking design promotes operator safety by securely covering the needle with an audible click
  • Help respond to the need for patient comfort and operator control
The Atellica VTR Arterial blood sampler syringes helps ensure simplicity, safety, and complete flexibility in sample collection. The ultra-sharp, thin-wall needle promotes smooth insertion, fast fill time, greater patient comfort, and sharps safety. The dual function, vented device allows the operator to choose either the preset plunger mode or manual aspiration. Additionally, the proven syringe vent system removes syringe dead space contamination, whether blood is drawn from an arterial puncture or arterial line.
All auto-venting syringes for direct sampling via arterial puncture come in 1 or 3 ml sizes and include a large venting filter for rapid filling with minimum back pressure. Auto-venting syringes are also available with a needle safety sheath.
Aspirating syringes sample directly from an arterial line and come in 1 or 3 ml sizes.
Packaging: 100 syringes per box, tripled-packaged, and filter caps are included.