Stainless Steel Buckets
Supplier: SAMPLING SYSTEMS LTD
A586-10 A586-12 A586-15
77578-044EA 322.28 USD
77578-044 77578-048 77578-052
Stainless Steel Buckets
Buckets
These general purpose buckets are ideal for use in all areas of industry.
  • Made from chemically resistant 316L stainless steel
  • Strong and robust heavy duty design
  • Perfect for corrosive environments
The range of stainless steel buckets from Sampling Systems are excellent multi purpose buckets that can be used across a wide range of industries. Made from chemically resistant 316L stainless, they are particularly suitable for use in corrosive environments.