Stirring Paddles, Stainless Steel
Supplier: SAMPLING SYSTEMS LTD
A344-1000 A344-1500 A916-1000 A916-1500
77577-882EA 162.77 USD
77577-882 77577-884 77578-162 77578-164
Stirring Paddles, Stainless Steel
Stirrers Stirring Tools
These paddles are ideal for use when producing liquids and suspensions. Stirrers are made from 316 stainless steel.
  • Robust construction
  • Crevice free
  • Satin finish
  • BSE/TSE free