Deep Water Samplers
Supplier: Sampling Systems
5079A-1000 5079A-2500
77576-538EA 811.63 USD
77576-538 77576-536
Deep Water Samplers
Samplers Water Samplers
The Deep water sampler can take samples of liquids from large depths.
  • Transparent plastic construction enables easy viewing
  • Ideal for sampling at large depths in rivers, lakes and large containers
  • Simple operation
The Deep water sampler is available in different sizes, it takes a sample at the point you start pulling the handle upwards. The simple operation means you lower to the required depth then pull it out.