Liquid Dip Samplers
Supplier: Sampling Systems
5175A-100 5175A-50
77576-558EA 274.91 USD
77576-558 77576-556
Liquid Dip Samplers
Samplers Sample Dippers
The stainless steel liquid dip sampler is used to collect a wide variety of liquid samples from the surface.
  • Made from high quality and chemically resistant 316L stainless steel
  • Different sizes available
  • Convenient hands-free emptying feature
Once the sample has been collected, the liquid dip sampler can simply be pushed against the container allowing for the liquid to be easily emptied.