GMP Pallet Knives
Supplier: SAMPLING SYSTEMS LTD
A123-100 A123-150 A123-200 A123-250 A123-300 A123-350
77577-634EA 132.29 USD
77577-634 77577-636 77577-638 77577-640 77577-642 77577-644
GMP Pallet Knives
Spatulas
This range of GMP pallet knives are excellent for cleaning, scraping and stirring.
  • Made from medical grade 420 stainless steel
  • Crevice-free and polished finish
  • Strong and flexible
  • Range of different lengths
The sampling systems range of stainless GMP pallet knives are ideal for a wide range of applications including cleaning, scraping and mixing. Made from medical grade 420 stainless steel they are crevice-free and have a polished finish meaning they are easily cleaned after use.