SteriWare® Powder Spatulas, Disposable
Supplier: SAMPLING SYSTEMS LTD
8095H-150 8095H-150S
77577-042EA 149.19 USD
77577-042 77577-044
SteriWare® Powder Spatulas, Disposable
Spatulas
HDPE, white. These disposable spatulas feature a rounded handle and a robust blade.
  • Manufactured and packed in a controlled atmosphere (class 100000 cleanroom)
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • Approved for use in ATEX Zones 0, 1 and 2 (contact Sampling Systems for full details)
  • Disposable: No cleaning is required before or after sampling
Suitable for sterile sampling and dispensing quantities of powders in all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.).