SteriWare® PowderSafe Powder Spatulas, Disposable
Supplier: SAMPLING SYSTEMS LTD
8097A-150 8097A-150S
77577-054EA 325.95 USD
77577-054 77577-056
SteriWare® PowderSafe Powder Spatulas, Disposable
Spatulas
PS, white. These disposable powder spatulas come complete with a click-on container.
  • Sampled powder is safely locked inside the cover
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • Manufactured in a medical cleanroom environment
  • Approved for use in ATEX Zones 0, 1 and 2
The transparent container slips over the blade of the spatula and is securely locked into place. At the laboratory the container can be easily removed. Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.).