SteriWare® Ladles, Disposable
Supplier: SAMPLING SYSTEMS LTD
8053B-50 8053B-50S 8053H-50 8053H-50S
77576-884EA 88.97 USD
77576-884 77576-886 77576-888 77576-890
SteriWare® Ladles, Disposable
Scoops
These disposable ladles are ideal for the sterile and non sterile surface sampling of liquids. The robust HDPE construction is ideal for a huge number of liquid and water testing applications.
  • Can be used straight-from-the-bag: Simply unwrap and use
  • Available in natural (HDPE) or blue (PP)
  • Blue version can be easily seen if it is left in the product by mistake
  • Moulded and packed in a class 100000 cleanroom
  • BSE/TSE-free
  • Full batch traceability
  • Available sterile (gamma irradiated)
The one piece moulded design has been specially constructed to go through 2" (51 mm) lid bung. 2" bungs are standard features on many barrels and kegs.
Certifications: Materials conform to FDA, EU 10/2011 and EC 1935/2004.
Packaging: Each sampler is individually bagged in a heat sealed PE bag.