SteriWare® Liquid Funnels
Supplier: SAMPLING SYSTEMS LTD
8370B-100 8370B-100S 8370H-100 8370H-100S 8370H-200 8370H-200S 8370H-50 8370H-50S
77577-328EA 83.06 USD
77577-328 77577-330 77577-332 77577-334 77577-336 77577-338 77577-340 77577-342
SteriWare® Liquid Funnels
Funnels Filtration Funnels Filter Funnels
Single use, disposable liquid funnels ideal for laboratories and manufacturing areas. Designed to use straight from bags.
  • Made of HDPE
  • Moulded and packed in a class 100,000 cleanroom
  • With small hygenic side handle
  • BSE/TSE free
  • Available sterile (gamma irradiated)
Certifications: Material conforms to FDA, EU 10/2011 and EC 1935/2004. Full batch traceability.
Packaging: Individually bagged.