SteriWare® Liquid and Powder Bottles
Supplier: Sampling Systems
SteriWare®
8404H-1000S 8404H-2000S 8404H-250S
77577-346EA 599.15 USD
77577-346 77577-348 77577-344
SteriWare® Liquid and Powder Bottles
Bottles Plastic Bottles
These sterile bottles are ideal for use in laboratories and production areas. The bottles have a wide neck which makes them ideal when working with powders as well as liquids.
  • Wide mouth bottles for easy filling
  • HDPE body with a screw on PP cap
  • Made from FDA conforming polymers
  • Packed in a Cleanroom
  • Individually bagged
  • Sterilised by Gamma
  • BSE/TSE-free
The packaging process takes place in a Class 7 Cleanroom and each bottle is packed into a laminate film bag. After packing the bottles are sterilised by gamma irradiation.