SteriWare® LiquiThief Disposable Samplers
Supplier: SAMPLING SYSTEMS LTD
8050H-1000 8050H-1000S 8050H-1500 8050H-1500S 8050H-500 8050H-500S 8050P-1000 8050P-500
77576-856EA 218.15 USD
77576-856 77576-858 77576-860 77576-862 77576-864 77576-866 77576-868 77576-870
SteriWare® LiquiThief Disposable Samplers
Samplers Disposable Samplers
Disposable liquid samplers designed for a high number of liquids and low viscosity testing applications.
  • No risk of cross- contamination - can be used straight from the bag
  • Silicone seal for improved sampling performance
  • Assembled and packed in a Class 100 000 cleanroom
Designed for taking fast sterile samples from tanks, kegs, and containers in pharmaceutical, food, chemical, cosmetic, and biotechnology industries.
Certifications: FDA, EU 10/2011 and EC 1935/2004 conformity. BSE-/TSE-free. Full batch traceability, ATEX rated.
Packaging: More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable