Steriware® Palette Knives, Disposable
Supplier: SAMPLING SYSTEMS LTD
8175H-150 8175H-150S 8175H-200 8175H-200S
77577-116EA 249.18 USD
77577-116 77577-118 77577-120 77577-122
Steriware® Palette Knives, Disposable
Spatulas
These disposable palette knives are single piece injection moulded from food/pharma grade HDPE. Ideal for use in manufacturing areas and laboratories.
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling
  • Manufactured in a class 100.000 cleanroom environment
  • Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.)
  • Available as gamma irradiated sterile (SAL 10⁻⁶)
  • BSE/TSE-free
FDA conforming material
Packaging: Individually bagged.