Conical Jugs, Graduated
Supplier: SAMPLING SYSTEMS LTD
A154-1000 A154-500
77577-712EA 33.95 USD
77577-712 77577-714
Conical Jugs, Graduated
Beakers
Graduated conical jugs for everyday use across a variety of applications.
  • Ideal for use in manufacturing, laboratories, etc
  • Robust and sturdy design
  • Easily visible graduations make measuring simple
  • Available from stock
  • Fast delivery
Stainless steel conical jugs feature easily visible graduation marks to make measuring simple. Their robust and sturdy design makes them easy to use and clean. They also feature an ergonomic handle.