La Crosse Wireless Indoor and Outdoor Weather Station
Supplier: LA CROSSE TECHNOLOGY
La Crosse
308-1414BV3
470356-812EA 69.95 USD
470356-812
La Crosse Wireless Indoor and Outdoor Weather Station
Weather Stations
This weather station provides precise, real-time weather data straight from the backyard or classroom.
  • 5 color forecast icons: sunny, partly sunny, cloudy, rainy, stormy
  • Outdoor transmission range up to 300 feet
  • Also includes heat index, dew point, atomic time and date, min./max. records
  • Display dimensions: 8.34"L×1.03"W×5.48"H
  • Sensor dimensions: 1.57"L×0.82"W×5.11"H
Indoor and outdoor temperature and humidity are conveniently displayed on the high contrast LCD display. Adjust the brightness when using the included power cord.
The animated weather forecast icons are determined by the station's internal barometric pressure reading. The forecast tendency arrows indicate at a glance if the pressure is rising, falling, or staying steady.
Ordering information: Display: 5.0 V AC adapter for primary power (included inside lower panel); requires 3 AAA Alkaline batteries for backup (not included). Sensor: powered by 2 AA Alkaline batteries (not included).