2-Ethynylaniline ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Aminophenylacetylene

E1410-250MG
TCE1410-250MGEA 38.62 USD
TCE1410-250MG
2-Ethynylaniline ≥97.0% (by GC, titration analysis)
2-Ethynylaniline
Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 101 °C
Flash Pt: 98 °C
MDL Number: MFCD00168862
CAS Number: 52670-38-9

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR