(Difluoromethyl)bis(2,5-dimethylphenyl)sulfonium tetrafluoroborate ≥95.0% (qNMR)

Supplier: TCI America
Danger

D5630-1G D5630-5G
TCD5630-1GEA 272 USD
TCD5630-1G TCD5630-5G
(Difluoromethyl)bis(2,5-dimethylphenyl)sulfonium tetrafluoroborate ≥95.0% (qNMR)
(Difluoromethyl)bis(2,5-dimethylphenyl)sulfonium tetrafluoroborate
Formula: C₁₇H₁₉BF₆S
CAS Number: 2133476-51-2
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (1H-qNMR) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR