2,7-Dibromospiro[9H-fluorene-9,9'-[9H]xanthene] ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,7-Dibromospiro[fluorene-9,9'-xanthene]

D5671-1G D5671-5G
TCD5671-1GEA 118 USD
TCD5671-1G TCD5671-5G
2,7-Dibromospiro[9H-fluorene-9,9'-[9H]xanthene] ≥98.0% (by GC)
2,7-Dibromospiro[9H-fluorene-9,9'-[9H]xanthene]
Formula: C₂₅H₁₄Br₂O
MDL Number: MFCD30342413
CAS Number: 198142-65-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR