Ethyl N-Cyanoacetimidate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: N-Cyanoethanimidic acid Ethyl ester, N-Cyanoacetimidic acid Ethyl ester, Ethyl N-Cyanoethanimidate, Ethyl N-cyanoacetimidate

E1331-25G E1331-5G
TCE1331-25GEA 98 USD
TCE1331-25G TCE1331-5G
Ethyl N-Cyanoacetimidate ≥98.0% (by GC)
Ethyl N-Cyanoacetimidate
Formula: C₅H₈N₂O
Boiling Pt: 96 °C
MDL Number: MFCD03411888
CAS Number: 1558-82-3
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR