1-Hexen-5-yne ≥97.0% (by GC)

Supplier: TCI America
Danger

H1541-1G H1541-5G
TCH1541-1GEA 275 USD
TCH1541-1G TCH1541-5G
1-Hexen-5-yne ≥97.0% (by GC)
1-Hexen-5-yne
Formula: C₆H₈
MDL Number: MFCD00060909
CAS Number: 14548-31-3
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR