4-Methoxy-N-methylbenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

M2836-1G M2836-5G
TCM2836-1GEA 40 USD
TCM2836-1G TCM2836-5G
4-Methoxy-N-methylbenzylamine ≥98.0% (by GC, titration analysis)
4-Methoxy-N-methylbenzylamine
Formula: C₉H₁₃NO
Boiling Pt: 238 °C
MDL Number: MFCD04115407
CAS Number: 702-24-9
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR