Dimethyl-2-aminoterephthalate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate

D5462-25G D5462-5G
TCD5462-25GEA 111 USD
TCD5462-25G TCD5462-5G
Dimethyl-2-aminoterephthalate ≥98.0% (by GC, titration analysis)
Dimethyl 2-aminoterephthalate
Formula: C₁₀H₁₁NO₄
MDL Number: MFCD00008427
CAS Number: 5372-81-6
EINECS: 226-364-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 133.0 to 137.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR