1,7-Dimethylxanthine ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
Paraxanthine, 1,7-Dimethyl-1H-purine-2,6(3H,7H)-dione
D5696-100MG
D5696-25MG
TCD5696-100MGEA
200
USD
TCD5696-100MG
TCD5696-25MG
1,7-Dimethylxanthine ≥98.0% (by HPLC)
1,7-Dimethylxanthine
Formula:
C₇H₈N₄O₂ MW: 180.17 |
MDL Number:
MFCD00005727 CAS Number: 611-59-6 EINECS: 210-271-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 30.50 to 31.70 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...