2-Iodobenzo[d]thiazole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Iodobenzothiazole

I0938-1G I0938-5G
TCI0938-1GEA 112 USD
TCI0938-1G TCI0938-5G
2-Iodobenzo[d]thiazole ≥98.0% (by GC)
2-Iodobenzo[d]thiazole
Formula: C₇H₄INS
MDL Number: MFCD00971971
CAS Number: 1123-99-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 81.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR