1-Bromodibenzofuran ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Bromodibenzo[b,d]furan

B5766-1G B5766-200MG
TCB5766-1GEA 204 USD
TCB5766-1G TCB5766-200MG
1-Bromodibenzofuran ≥98.0% (by GC)
1-Bromodibenzofuran
Formula: C₁₂H₇BrO
MDL Number: MFCD00094238
CAS Number: 50548-45-3

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR