Chlorotrimethoxysilane ≥90.0% (by GC)

Supplier: TCI America
Danger

Synonyms: Trimethoxychlorosilane

C3617-25G C3617-5G
TCC3617-25GEA 187 USD
TCC3617-25G TCC3617-5G
Chlorotrimethoxysilane ≥90.0% (by GC)
Chlorotrimethoxysilane
Formula: C₃H₉ClO₃Si
MW: 156.64 g/mol
Boiling Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191934
CAS Number: 4668-00-2
UN: 2985
ADR: 3 II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR