3,7-Dibromo-10-hexylphenothiazine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
D5222-1G
D5222-5G
TCD5222-1GEA
163
USD
TCD5222-1G
TCD5222-5G
3,7-Dibromo-10-hexylphenothiazine ≥98.0% (by HPLC, total nitrogen)
3,7-Dibromo-10-hexylphenothiazine
Formula:
C₁₈H₁₉Br₂NS |
CAS Number:
312924-93-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 54.0 to 58.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...