3-(2-Amino-2-oxoethyl)-5-methylhexanoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
3-(Carbamoylmethyl)-5-methylhexanoic acid
A3287-100G
A3287-25G
TCA3287-100GEA
58
USD
TCA3287-100G
TCA3287-25G
3-(2-Amino-2-oxoethyl)-5-methylhexanoic acid ≥98.0% (by HPLC, titration analysis)
3-(2-Amino-2-oxoethyl)-5-methylhexanoic acid
Formula:
C₉H₁₇NO₃ MW: 187.24 g/mol |
MDL Number:
MFCD08457698 CAS Number: 181289-15-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 132.0 to 136.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...