10-Acetylphenothiazine ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1-(10H-Phenothiazin-10-yl)ethanone, 10-Acetyl-10H-phenothiazine
A3154-1G
A3154-5G
TCA3154-1GEA
117
USD
TCA3154-1G
TCA3154-5G
10-Acetylphenothiazine ≥98.0% (by GC)
10-Acetylphenothiazine
Formula:
C₁₄H₁₁NOS MW: 241.31 g/mol |
MDL Number:
MFCD00022264 CAS Number: 1628-29-1 EINECS: 216-621-7 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 202.0 to 206.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...