4-(9H-Carbazol-9-yl)benzaldehyde ≥96.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
9-(4-Formylphenyl)-9H-carbazole
C3481-1G
C3481-5G
TCC3481-1GEA
85
USD
TCC3481-1G
TCC3481-5G
4-(9H-Carbazol-9-yl)benzaldehyde ≥96.0% (by GC)
4-(9H-Carbazol-9-yl)benzaldehyde
Formula:
C₁₉H₁₃NO MW: 271.32 g/mol |
MDL Number:
MFCD19441374 CAS Number: 110677-45-7 |
Specification Test Results
Appearance | White to Amber powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 156.0 to 160.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...