Chloro(3-chloropropyl)dimethylsilane ≥96.0% (by GC)

Supplier: TCI America
Danger

C3426-1ML C3426-5ML
TCC3426-1MLEA 71 USD
TCC3426-1ML TCC3426-5ML
Chloro(3-chloropropyl)dimethylsilane ≥96.0% (by GC)
Chloro(3-chloropropyl)dimethylsilane
Formula: C₅H₁₂Cl₂Si
Boiling Pt: 179 °C
MDL Number: MFCD00042612
CAS Number: 10605-40-0
EINECS: 234-234-1
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR